Anti-FGF6 Mouse Monoclonal Antibody [clone: 27028]

Supplier: R & D Systems
MAB238
220019-811EA 763.57 USD
220019-811
Anti-FGF6 Mouse Monoclonal Antibody [clone: 27028]
Antibodies
Type: Primary
Antigen Symbol: FGF6
Antigen Name: fibroblast growth factor 6
Antigen Synonyms: HBGF-6, HST2
Clonality: Monoclonal
Clone: 27028
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR