Anti-CD93 Mouse Monoclonal Antibody [clone: 273107]

Supplier: R & D Systems
MAB23791
220019-810EA 594.37 USD
220019-810
Anti-CD93 Mouse Monoclonal Antibody [clone: 273107]
Antibodies
Type: Primary
Antigen: CD93
Clonality: Monoclonal
Clone: 273107
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR