Anti-TNFRSF23 Rat Monoclonal Antibody [clone: 270028]

Supplier: R & D Systems
MAB23781
220019-809EA 577.02 USD
220019-809
Anti-TNFRSF23 Rat Monoclonal Antibody [clone: 270028]
Antibodies
Type: Primary
Antigen: TNFRSF23
Clonality: Monoclonal
Clone: 270028
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR