Anti-AMIGO3 Rat Monoclonal Antibody [clone: 37131]

Supplier: R & D Systems
MAB2375
220019-806EA 594.37 USD
220019-806
Anti-AMIGO3 Rat Monoclonal Antibody [clone: 37131]
Antibodies
Type: Primary
Antigen: AMIGO3
Clonality: Monoclonal
Clone: 37131
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR