Anti-AMIGO2 Rat Monoclonal Antibody [clone: 29728]

Supplier: R & D Systems
MAB2374
220019-805EA 577.02 USD
220019-805
Anti-AMIGO2 Rat Monoclonal Antibody [clone: 29728]
Antibodies
Type: Primary
Antigen: AMIGO2
Clonality: Monoclonal
Clone: 29728
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR