Anti-FGF5 Mouse Monoclonal Antibody [clone: 221907]

Supplier: R & D Systems
MAB237
220019-802EA 772.25 USD
220019-802
Anti-FGF5 Mouse Monoclonal Antibody [clone: 221907]
Antibodies
Type: Primary
Antigen Symbol: FGF5
Antigen Name: fibroblast growth factor 5
Antigen Synonyms: HBGF-5, Smag-82, TCMGLY
Clonality: Monoclonal
Clone: 221907
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR