Anti-HSPG2 Mouse Monoclonal Antibody [clone: 268908]

Supplier: R & D Systems
MAB2364
220019-799EA 594.37 USD
220019-799
Anti-HSPG2 Mouse Monoclonal Antibody [clone: 268908]
Antibodies
Type: Primary
Antigen: HSPG2
Clonality: Monoclonal
Clone: 268908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR