Anti-DPP6 Mouse Monoclonal Antibody [clone: 274311]

Supplier: R & D Systems
MAB23601
220019-798EA 577.02 USD
220019-798
Anti-DPP6 Mouse Monoclonal Antibody [clone: 274311]
Antibodies
Type: Primary
Antigen: DPP6
Clonality: Monoclonal
Clone: 274311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR