Anti-CD69 Mouse Monoclonal Antibody [clone: 298633]

Supplier: R & D Systems
MAB2359
220019-794EA 594.37 USD
220019-794
Anti-CD69 Mouse Monoclonal Antibody [clone: 298633]
Antibodies
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: 298633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR