Anti-MCFD2 Mouse Monoclonal Antibody [clone: 294301]

Supplier: R & D Systems
MAB2357
220019-793EA 561.11 USD
220019-793
Anti-MCFD2 Mouse Monoclonal Antibody [clone: 294301]
Antibodies
Type: Primary
Antigen: MCFD2
Clonality: Monoclonal
Clone: 294301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR