Anti-GPC2 Rat Monoclonal Antibody [clone: 300712]

Supplier: R & D Systems
MAB23551
220019-792EA 566.89 USD
220019-792
Anti-GPC2 Rat Monoclonal Antibody [clone: 300712]
Antibodies
Type: Primary
Antigen Symbol: GPC2
Antigen Name: glypican 2 (cerebroglycan)
Antigen Synonyms: 2410016G05Rik
Clonality: Monoclonal
Clone: 300712
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR