Anti-CCRL2 Mouse Monoclonal Antibody [clone: 152211]

Supplier: R & D Systems
MAB2350
220019-789EA 763.57 USD
220019-789
Anti-CCRL2 Mouse Monoclonal Antibody [clone: 152211]
Antibodies
Type: Primary
Antigen: CCRL2
Clonality: Monoclonal
Clone: 152211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR