Anti-FGF4 Mouse Monoclonal Antibody [clone: 19805]

Supplier: R & D Systems
MAB235
220019-788EA 763.57 USD
220019-788
Anti-FGF4 Mouse Monoclonal Antibody [clone: 19805]
Antibodies
Type: Primary
Antigen Symbol: FGF4
Antigen Name: fibroblast growth factor 4
Antigen Synonyms: HBGF-4, HST, HST-1, HSTF1, K-FGF, KFGF
Clonality: Monoclonal
Clone: 19805
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR