Anti-VTN Mouse Monoclonal Antibody [clone: 342603]

Supplier: R & D Systems
MAB2349
220019-787EA 633.42 USD
220019-787
Anti-VTN Mouse Monoclonal Antibody [clone: 342603]
Antibodies
Simple Western

Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone: 342603
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR