Anti-SPOCK3 Rat Monoclonal Antibody [clone: 330404]

Supplier: R & D Systems
MAB23461
220019-786EA 561.11 USD
220019-786
Anti-SPOCK3 Rat Monoclonal Antibody [clone: 330404]
Antibodies
Type: Primary
Antigen: SPOCK3
Clonality: Monoclonal
Clone: 330404
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR