Anti-SPOCK3 Rat Monoclonal Antibody [clone: 330403]

Supplier: R & D Systems
MAB2346
220019-785EA 763.57 USD
220019-785
Anti-SPOCK3 Rat Monoclonal Antibody [clone: 330403]
Antibodies
Type: Primary
Antigen: SPOCK3
Clonality: Monoclonal
Clone: 330403
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR