Anti-CAR12 Rat Monoclonal Antibody [clone: 336103]

Supplier: R & D Systems
MAB2345
220019-784EA 561.11 USD
220019-784
Anti-CAR12 Rat Monoclonal Antibody [clone: 336103]
Antibodies
Type: Primary
Antigen: CAR12
Clonality: Monoclonal
Clone: 336103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR