Anti-MMEL1 Mouse Monoclonal Antibody [clone: 353207]

Supplier: R & D Systems
MAB2340
220019-783EA 577.02 USD
220019-783
Anti-MMEL1 Mouse Monoclonal Antibody [clone: 353207]
Antibodies
Type: Primary
Antigen: MMEL1
Clonality: Monoclonal
Clone: 353207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR