Anti-F3 Mouse Monoclonal Antibody [clone: 323519]

Supplier: R & D Systems
MAB23391
220019-780EA 584.25 USD
220019-780
Anti-F3 Mouse Monoclonal Antibody [clone: 323519]
Antibodies
Type: Primary
Antigen: F3
Clonality: Monoclonal
Clone: 323519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR