Anti-F3 Mouse Monoclonal Antibody [clone: 323514]

Supplier: R & D Systems
MAB2339-500 MAB2339-100
220019-782EA 1459.17 USD
220019-782 220019-781
Anti-F3 Mouse Monoclonal Antibody [clone: 323514]
Antibodies
Type: Primary
Antigen: F3
Clonality: Monoclonal
Clone: 323514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR