Anti-F7 Mouse Monoclonal Antibody [clone: 321605]

Supplier: R & D Systems
MAB23381
220019-779EA 584.25 USD
220019-779
Anti-F7 Mouse Monoclonal Antibody [clone: 321605]
Antibodies
Type: Primary
Antigen Symbol: F7
Antigen Name: coagulation factor VII (serum prothrombin conversion accelerator)
Antigen Synonyms: SPCA
Clonality: Monoclonal
Clone: 321605
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR