Anti-F7 Mouse Monoclonal Antibody [clone: 321621]

Supplier: R & D Systems
MAB2338
220019-778EA 633.42 USD
220019-778
Anti-F7 Mouse Monoclonal Antibody [clone: 321621]
Antibodies
Type: Primary
Antigen Symbol: F7
Antigen Name: coagulation factor VII (serum prothrombin conversion accelerator)
Antigen Synonyms: SPCA
Clonality: Monoclonal
Clone: 321621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR