Anti-GPNMB Rat Monoclonal Antibody [clone: 297310]

Supplier: R & D Systems
MAB2330
220019-775EA 594.37 USD
220019-775
Anti-GPNMB Rat Monoclonal Antibody [clone: 297310]
Antibodies
Type: Primary
Antigen Symbol: GPNMB
Antigen Name: glycoprotein (transmembrane) nmb
Antigen Synonyms: DC-HIL, Dchil, ipd
Clonality: Monoclonal
Clone: 297310
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR