Anti-SPOCK2 Mouse Monoclonal Antibody [clone: 327403]

Supplier: R & D Systems
MAB2328
220019-773EA 566.89 USD
220019-773
Anti-SPOCK2 Mouse Monoclonal Antibody [clone: 327403]
Antibodies
Type: Primary
Antigen: SPOCK2
Clonality: Monoclonal
Clone: 327403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR