Anti-SPOCK1 Mouse Monoclonal Antibody [clone: 316801]

Supplier: R & D Systems
MAB2327
220019-772EA 577.02 USD
220019-772
Anti-SPOCK1 Mouse Monoclonal Antibody [clone: 316801]
Antibodies
Type: Primary
Antigen: SPOCK1
Clonality: Monoclonal
Clone: 316801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR