Anti-RARRES2 Rat Monoclonal Antibody [clone: 372409]

Supplier: R & D Systems
MAB23251
220019-771EA 763.57 USD
220019-771
Anti-RARRES2 Rat Monoclonal Antibody [clone: 372409]
Antibodies
Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 372409
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR