Anti-RARRES2 Mouse Monoclonal Antibody [clone: 365317]

Supplier: R & D Systems
MAB2324
220019-768EA 594.37 USD
220019-768
Anti-RARRES2 Mouse Monoclonal Antibody [clone: 365317]
Antibodies
Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 365317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR