Anti-LAG3 Mouse Monoclonal Antibody [clone: 874501]

Supplier: R & D Systems
MAB23193
102885-454EA 655.11 USD
102885-454
Anti-LAG3 Mouse Monoclonal Antibody [clone: 874501]
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 874501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR