Anti-LAG3 Mouse Monoclonal Antibody [clone: 333210]

Supplier: R & D Systems
MAB2319
220019-764EA 633.42 USD
220019-764
Anti-LAG3 Mouse Monoclonal Antibody [clone: 333210]
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 333210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR