Anti-INPP5D Rat Monoclonal Antibody [clone: 257812]

Supplier: R & D Systems
MAB2317
220019-763EA 584.25 USD
220019-763
Anti-INPP5D Rat Monoclonal Antibody [clone: 257812]
Antibodies
Type: Primary
Antigen: INPP5D
Clonality: Monoclonal
Clone: 257812
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR