Anti-AKT2 Mouse Monoclonal Antibody [clone: 3251]

Supplier: R & D Systems
MAB23152
220019-762EA 577.02 USD
220019-762
Anti-AKT2 Mouse Monoclonal Antibody [clone: 3251]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 3251
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR