Anti-HSPB1 (phospho-Ser78/S82) Rabbit Monoclonal Antibody [clone: 1026B]

Supplier: R & D Systems
MAB23141
102885-452EA 600.15 USD
102885-452
Anti-HSPB1 (phospho-Ser78/S82) Rabbit Monoclonal Antibody [clone: 1026B]
Antibodies
Type: Primary
Antigen Symbol: HSPB1
Antigen Name: heat shock 27kDa protein 1
Antigen Synonyms: CMT2F, HEL-S-102, HMN2B, HS.76067, HSP27, HSP28, Hsp25, SRP27
Clonality: Monoclonal
Clone: 1026B
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR