Anti-MBL2 Mouse Monoclonal Antibody [clone: 285623]

Supplier: R & D Systems
MAB23071
220019-759EA 772.25 USD
220019-759
Anti-MBL2 Mouse Monoclonal Antibody [clone: 285623]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: MBL2
Clonality: Monoclonal
Clone: 285623
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR