Anti-MBL2 Mouse Monoclonal Antibody [clone: 285602]

Supplier: R & D Systems
MAB2307
220019-758EA 594.37 USD
220019-758
Anti-MBL2 Mouse Monoclonal Antibody [clone: 285602]
Antibodies
Type: Primary
Antigen: MBL2
Clonality: Monoclonal
Clone: 285602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR