Anti-IL17C Rat Monoclonal Antibody [clone: 311522]

Supplier: R & D Systems
MAB23061
220019-757EA 566.89 USD
220019-757
Anti-IL17C Rat Monoclonal Antibody [clone: 311522]
Antibodies
Type: Primary
Antigen: IL17C
Clonality: Monoclonal
Clone: 311522
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR