Anti-IL17C Rat Monoclonal Antibody [clone: 311523]

Supplier: R & D Systems
MAB2306
220019-756EA 608.83 USD
220019-756
Anti-IL17C Rat Monoclonal Antibody [clone: 311523]
Antibodies
Type: Primary
Antigen: IL17C
Clonality: Monoclonal
Clone: 311523
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR