Anti-IL6 Mouse Monoclonal Antibody [clone: 341031]

Supplier: R & D Systems
MAB23051
220019-755EA 561.11 USD
220019-755
Anti-IL6 Mouse Monoclonal Antibody [clone: 341031]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 341031
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR