Anti-IL6 Mouse Monoclonal Antibody [clone: 341041]

Supplier: R & D Systems
MAB2305
220019-754EA 772.25 USD
220019-754
Anti-IL6 Mouse Monoclonal Antibody [clone: 341041]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 341041
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR