Anti-IFNG Rat Monoclonal Antibody [clone: 345001]

Supplier: R & D Systems
MAB23001
220019-752EA 772.25 USD
220019-752
Anti-IFNG Rat Monoclonal Antibody [clone: 345001]
Antibodies
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 345001
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR