Anti-IFNG Rat Monoclonal Antibody [clone: 345025]

Supplier: R & D Systems
MAB2300
220019-751EA 577.02 USD
220019-751
Anti-IFNG Rat Monoclonal Antibody [clone: 345025]
Antibodies
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 345025
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR