Anti-BMPER Rat Monoclonal Antibody [clone: 349920]

Supplier: R & D Systems
MAB2299
220019-749EA 561.11 USD
220019-749
Anti-BMPER Rat Monoclonal Antibody [clone: 349920]
Antibodies
Type: Primary
Antigen: BMPER
Clonality: Monoclonal
Clone: 349920
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR