Anti-IL1F6 Rat Monoclonal Antibody [clone: 275339]

Supplier: R & D Systems
MAB2297
220019-747EA 763.57 USD
220019-747
Anti-IL1F6 Rat Monoclonal Antibody [clone: 275339]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: IL1F6
Clonality: Monoclonal
Clone: 275339
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR