Anti-CD22 Rat Monoclonal Antibody [clone: 308501]

Supplier: R & D Systems
MAB2296
220019-746EA 594.37 USD
220019-746
Anti-CD22 Rat Monoclonal Antibody [clone: 308501]
Antibodies
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: 308501
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR