Anti-TNFSF9 Mouse Monoclonal Antibody [clone: 282220]

Supplier: R & D Systems
MAB2295
220019-745EA 594.37 USD
220019-745
Anti-TNFSF9 Mouse Monoclonal Antibody [clone: 282220]
Antibodies
Type: Primary
Antigen: TNFSF9
Clonality: Monoclonal
Clone: 282220
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR