Anti-PIN1 Mouse Monoclonal Antibody [clone: 257417]

Supplier: R & D Systems
MAB2294
220019-744EA 639.2 USD
220019-744
Anti-PIN1 Mouse Monoclonal Antibody [clone: 257417]
Antibodies
Type: Primary
Antigen: PIN1
Clonality: Monoclonal
Clone: 257417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR