Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 81316]

Supplier: R & D Systems
MAB2291
220019-743EA 786.71 USD
220019-743
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 81316]
Antibodies
Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 81316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR