Anti-ATM Mouse Monoclonal Antibody [clone: 664703]

Supplier: R & D Systems
MAB22901
102885-450EA 577.02 USD
102885-450
Anti-ATM Mouse Monoclonal Antibody [clone: 664703]
Antibodies
Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 664703
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR