Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 81337]

Supplier: R & D Systems
MAB229
220019-742EA 779.48 USD
220019-742
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 81337]
Antibodies
Agonist Activity

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 81337
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR