Anti-CHEK2 Rat Monoclonal Antibody [clone: 235303]

Supplier: R & D Systems
MAB22851
220019-740EA 566.89 USD
220019-740
Anti-CHEK2 Rat Monoclonal Antibody [clone: 235303]
Antibodies
Type: Primary
Antigen: CHEK2
Clonality: Monoclonal
Clone: 235303
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR