Anti-CXCL8 Rat Monoclonal Antibody [clone: 344615]

Supplier: R & D Systems
MAB22772
220019-736EA 763.57 USD
220019-736
Anti-CXCL8 Rat Monoclonal Antibody [clone: 344615]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 344615
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR